siRNA Details
| Gene Symbol: Asgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012503.2 | NP_036635.1 | 7, 8 | 661 | ||
| XM_006246579.1 | XP_006246641.1 | 7, 8 | 773 | |||
| XM_008767768.1 | XP_008765990.1 | 7, 8 | 793 | |||
| XM_008767769.1 | XP_008765991.1 | 7, 8 | 737 | |||
| GenBank mRNA | BC088154.1 | - | 7, 8 | 661 | 7372227 | |
| FQ209763.1 | - | 7, 8 | 697 | |||
| FQ218553.1 | - | 7, 8 | 737 | |||
| FQ218644.1 | - | 7, 8 | 697 | |||
| FQ218824.1 | - | 7, 8 | 697 | |||
| FQ219107.1 | - | 7, 8 | 734 | |||
| FQ219710.1 | - | 7, 8 | 778 | |||
| M21770.1 | - | 6, 7 | 563 | |||

Asgr1
Genomic Map