siRNA Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012504.1 | NP_036636.1 | 4 | 578 | ||
| GenBank mRNA | BC061968.1 | - | 4 | 589 | 5598014 | |
| M14511.1 | - | 4 | 578 | |||
| M28647.1 | - | 4 | 601 | |||
| X05882.1 | - | 4 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1a1| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012504.1 | NP_036636.1 | 4 | 578 | ||
| GenBank mRNA | BC061968.1 | - | 4 | 589 | 5598014 | |
| M14511.1 | - | 4 | 578 | |||
| M28647.1 | - | 4 | 601 | |||
| X05882.1 | - | 4 | 355 | |||