siRNA Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012506.1 | NP_036638.1 | 11 | 1520 | ||
| XM_008758888.1 | XP_008757110.1 | 11 | 1434 | |||
| GenBank mRNA | FQ212331.1 | - | 10 | 1357 | ||
| M14513.1 | - | 11 | 1520 | |||
| M28648.1 | - | 10 | 1374 | |||
| X05883.1 | - | 11 | 1517 | |||

Atp1a3
Genomic Map