siRNA Details
| Gene Symbol: Atp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012509.1 | NP_036641.1 | 11 | 1832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp4a| Gene Symbol: Atp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012509.1 | NP_036641.1 | 11 | 1832 | ||