siRNA Details
| Gene Symbol: C4bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016995.2 | NP_058691.2 | 4 | 711 | ||
| XM_006249705.2 | XP_006249767.1 | 5 | 767 | |||
| XM_008769423.1 | XP_008767645.1 | 5 | 584 | |||
| GenBank mRNA | BC088152.1 | - | 4 | 711 | 7322205 | |
| FQ209636.1 | - | 4 | 722 | |||
| FQ218474.1 | - | 4 | 692 | |||
| Z50052.1 | - | 4 | 690 | |||

C4bpb
Genomic Map