siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 10 | 1575 | ||
| XM_008760747.1 | XP_008758969.1 | 11 | 1760 | |||
| XM_008760749.1 | XP_008758971.1 | 11 | 1745 | |||
| XM_008760750.1 | XP_008758972.1 | 11 | 1763 | |||
| GenBank mRNA | AY230250.1 | - | 10 | 1577 | ||

C6
Genomic Map