siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012517.2 | NP_036649.2 | 29 | 4033 | ||
XM_006237159.2 | XP_006237221.1 | 31 | 3992 | |||
XM_006237170.2 | XP_006237232.1 | 29 | 3878 | |||
XM_006237171.2 | XP_006237233.1 | 29 | 3878 | |||
XM_006237172.2 | XP_006237234.1 | 29 | 3878 | |||
XM_006237175.2 | XP_006237237.1 | 30 | 4386 | |||
XM_008763216.1 | XP_008761438.1 | 30 | 5895 | |||
GenBank mRNA | AF394938.1 | - | 29 | 3884 | ||
AF394939.1 | - | 29 | 3884 | |||
AF394940.1 | - | 29 | 3884 | |||
AY974797.2 | - | 31 | 4150 | |||
DQ538522.1 | - | 31 | 3852 | |||
M59786.1 | - | 29 | 4033 | |||
M67515.1 | - | 29 | 4027 | |||
M67516.1 | - | 29 | 4512 | |||
S80558.1 | - | 2 | 152 | |||
U31815.1 | - | 3 | 243 |
