siRNA Details
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016996.1 | NP_058692.1 | 4 | 1274 | ||
GenBank mRNA | AY214122.1 | - | 4 | 1264 | ||
U10354.1 | - | 4 | 1274 | |||
U20289.1 | - | 3 | 900 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
