siRNA Details
| Gene Symbol: Cbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012522.2 | NP_036654.2 | 2 | 262 | ||
| GenBank mRNA | D01098.1 | - | 2 | 263 | ||
| FQ211441.1 | - | 2 | 321 | |||
| M88344.1 | - | 2 | 411 | |||
| M88346.1 | - | 1 | 157 | |||
| M88347.1 | - | 1 | 157 | |||

Cbs
Genomic Map