siRNA Details
| Gene Symbol: Cbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012522.2 | NP_036654.2 | 6 | 756 | ||
| GenBank mRNA | D01098.1 | - | 6 | 757 | ||
| FQ211441.1 | - | 6 | 815 | |||
| M88344.1 | - | 6 | 905 | |||
| M88345.1 | - | 3 | 1169 | |||
| M88346.1 | - | 5 | 651 | |||
| M88347.1 | - | 5 | 651 | |||

Cbs
Genomic Map