siRNA Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031506.1 | NP_113694.1 | 10 | 1412 | ||
| XM_006236097.2 | XP_006236159.1 | 11 | 1531 | |||
| XM_008762680.1 | XP_008760902.1 | 11 | 1531 | |||
| GenBank mRNA | FJ959076.1 | - | 10 | 1170 | ||

Cftr
Genomic Map