siRNA Details
| Gene Symbol: Chgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012526.2 | NP_036658.2 | 4 | 1749 | ||
| XM_006235072.2 | XP_006235134.1 | 5 | 1659 | |||
| GenBank mRNA | AF019974.1 | - | 4 | 1549 | ||
| BC128743.1 | - | 4 | 1568 | 8370781 | ||

Chgb
Genomic Map