siRNA Details
| Gene Symbol: Chrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012527.2 | NP_036659.2 | 5 | 668 | ||
| XM_008771690.1 | XP_008769912.1 | 6 | 614 | |||
| GenBank mRNA | AB017656.1 | - | 1 | 121 | ||
| M16407.1 | - | 2 | 161 | |||
| M18088.1 | - | 3 | 252 | |||
More Information
Set Membership: |
|

Chrm3
Genomic Map