siRNA Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012528.1 | NP_036660.1 | 7 | 854 | ||
| XM_008767770.1 | XP_008765992.1 | 7 | 901 | |||
| GenBank mRNA | X74833.1 | - | 7 | 854 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnb1| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012528.1 | NP_036660.1 | 7 | 854 | ||
| XM_008767770.1 | XP_008765992.1 | 7 | 901 | |||
| GenBank mRNA | X74833.1 | - | 7 | 854 | ||