siRNA Details
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012528.1 | NP_036660.1 | 9 | 1178 | ||
XM_008767770.1 | XP_008765992.1 | 9 | 1225 | |||
GenBank mRNA | X74833.1 | - | 9 | 1178 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012528.1 | NP_036660.1 | 9 | 1178 | ||
XM_008767770.1 | XP_008765992.1 | 9 | 1225 | |||
GenBank mRNA | X74833.1 | - | 9 | 1178 |