siRNA Details
| Gene Symbol: Cyp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012541.3 | NP_036673.3 | 6 | 1257 | ||
| GenBank mRNA | BC127476.1 | - | 6 | 1257 | 7373253 | |
| FQ209607.1 | - | 5 | 685 | |||
| FQ210213.1 | - | 6 | 1263 | |||
| FQ210219.1 | - | 5 | 405 | |||
| K02422.1 | - | 5 | 1225 | |||
| X01031.1 | - | 5 | 772 | |||

Cyp1a2
Genomic Map