siRNA Details
| Gene Symbol: Cyp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012541.3 | NP_036673.3 | 4 | 1061 | ||
| GenBank mRNA | BC127476.1 | - | 4 | 1061 | 7373253 | |
| FQ209607.1 | - | 3 | 489 | |||
| FQ210213.1 | - | 4 | 1067 | |||
| FQ210219.1 | - | 3 | 209 | |||
| K02422.1 | - | 3 | 1029 | |||
| M26127.1 | - | 3 | 218 | |||
| X01031.1 | - | 3 | 576 | |||

Cyp1a2
Genomic Map