siRNA Details
| Gene Symbol: Cyp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012541.3 | NP_036673.3 | 2 | 62 | ||
| GenBank mRNA | BC127476.1 | - | 2 | 62 | 7373253 | |
| FQ210213.1 | - | 2 | 68 | |||
| K02422.1 | - | 1 | 30 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp1a2| Gene Symbol: Cyp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012541.3 | NP_036673.3 | 2 | 62 | ||
| GenBank mRNA | BC127476.1 | - | 2 | 62 | 7373253 | |
| FQ210213.1 | - | 2 | 68 | |||
| K02422.1 | - | 1 | 30 | |||