siRNA Details
| Gene Symbol: Nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017000.3 | NP_058696.2 | 3 | 345 | ||
| GenBank mRNA | BC083542.1 | - | 3 | 345 | 7190779 | |
| J02608.1 | - | 3 | 330 | |||
| J02640.1 | - | 2 | 304 | |||
| J02679.1 | - | 3 | 330 | |||
| X17464.1 | - | 1 | 19 | |||

Nqo1
Genomic Map