siRNA Details
| Gene Symbol: Drd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253599.2 | XP_006253661.1 | 2 | 1010 | ||
| XM_006253600.2 | XP_006253662.1 | 2 | 823 | |||
| GenBank mRNA | M35077.1 | - | 1 | 146 | ||
More Information
Set Membership: |
|

Drd1
Genomic Map