siRNA Details
| Gene Symbol: Ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012555.2 | NP_036687.1 | 3 | 568 | ||
| XM_006242752.1 | XP_006242814.1 | 3 | 616 | |||
| GenBank mRNA | BC101927.1 | - | 3 | 606 | 7939228 | |
| L20681.1 | - | 3 | 583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ets1| Gene Symbol: Ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012555.2 | NP_036687.1 | 3 | 568 | ||
| XM_006242752.1 | XP_006242814.1 | 3 | 616 | |||
| GenBank mRNA | BC101927.1 | - | 3 | 606 | 7939228 | |
| L20681.1 | - | 3 | 583 | |||