siRNA Details
Gene Symbol: Ets1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012555.2 | NP_036687.1 | 1 | 305 | ||
XM_006242752.1 | XP_006242814.1 | 1 | 353 | |||
GenBank mRNA | BC101927.1 | - | 1 | 343 | 7939228 | |
L20681.1 | - | 1 | 320 |
