siRNA Details
| Gene Symbol: Fbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012558.3 | NP_036690.2 | 2 | 517 | ||
| GenBank mRNA | BC078894.1 | - | 2 | 499 | 7114766 | |
| BC078895.1 | - | 2 | 517 | 7134943 | ||
| J04112.1 | - | 2 | 313 | |||
| M86240.1 | - | 2 | 403 | |||

Fbp1
Genomic Map