siRNA Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020071.2 | NP_064456.2 | 8 | 1436 | ||
XM_006232524.2 | XP_006232586.1 | 8 | 1454 | |||
GenBank mRNA | BC088102.1 | - | 8 | 1436 | 7371665 | |
FQ209924.1 | - | 8 | 1443 | |||
FQ210796.1 | - | 2 | 360 | |||
FQ218488.1 | - | 3 | 520 | |||
FQ219024.1 | - | 8 | 1348 | |||
M27220.1 | - | 5 | 848 | |||
U05675.1 | - | 8 | 1437 |
