siRNA Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012559.2 | NP_036691.2 | 3 | 185 | ||
| XM_006232525.1 | XP_006232587.1 | 3 | 339 | |||
| GenBank mRNA | BC078893.1 | - | 3 | 185 | 7122366 | |
| FQ219408.1 | - | 3 | 205 | |||
| FQ219511.1 | - | 3 | 207 | |||
| FQ219541.1 | - | 3 | 207 | |||
| FQ219604.1 | - | 3 | 206 | |||
| FQ229132.1 | - | 3 | 212 | |||
| J00734.1 | - | 3 | 170 | |||

Fgg
Genomic Map