siRNA Details
| Gene Symbol: Fh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017005.2 | NP_058701.2 | 9 | 1356 | ||
| GenBank mRNA | BC087598.1 | - | 9 | 1356 | 7311498 | |
| FQ218764.1 | - | 5 | 667 | |||
| FQ230216.1 | - | 5 | 701 | |||
| FQ232207.1 | - | 5 | 701 | |||
| J04473.1 | - | 9 | 1367 | |||

Fh
Genomic Map