siRNA Details
| Gene Symbol: Fh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017005.2 | NP_058701.2 | 10 | 1438 | ||
| GenBank mRNA | BC087598.1 | - | 10 | 1438 | 7311498 | |
| FQ218764.1 | - | 6 | 749 | |||
| FQ230216.1 | - | 6 | 783 | |||
| FQ232207.1 | - | 6 | 783 | |||
| J04473.1 | - | 10 | 1449 | |||

Fh
Genomic Map