siRNA Details
| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012563.1 | NP_036695.1 | 5 | 652 | ||
| GenBank mRNA | M72422.1 | - | 5 | 652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gad2| Gene Symbol: Gad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012563.1 | NP_036695.1 | 5 | 652 | ||
| GenBank mRNA | M72422.1 | - | 5 | 652 | ||