siRNA Details
| Gene Symbol: Gls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109968.1 | NP_001103438.1 | 5 | 859 | ||
| NM_012569.2 | NP_036701.2 | 5 | 859 | |||
| XM_008767075.1 | XP_008765297.1 | 5 | 1030 | |||
| GenBank mRNA | M65150.1 | - | 5 | 866 | ||

Gls
Genomic Map