siRNA Details
| Gene Symbol: Gls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109968.1 | NP_001103438.1 | 7 | 1085 | ||
| NM_012569.2 | NP_036701.2 | 7 | 1085 | |||
| XM_008767075.1 | XP_008765297.1 | 7 | 1256 | |||
| GenBank mRNA | AY083459.1 | - | 2 | 140 | ||
| M65150.1 | - | 7 | 1092 | |||

Gls
Genomic Map