siRNA Details
| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012570.2 | NP_036702.1 | 7 | 1184 | ||
| GenBank mRNA | BC081841.1 | - | 7 | 1068 | 7107138 | |
| X14044.1 | - | 7 | 1074 | |||
| X14223.1 | - | 7 | 1047 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glud1| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012570.2 | NP_036702.1 | 7 | 1184 | ||
| GenBank mRNA | BC081841.1 | - | 7 | 1068 | 7107138 | |
| X14044.1 | - | 7 | 1074 | |||
| X14223.1 | - | 7 | 1047 | |||