siRNA Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012571.2 | NP_036703.2 | 4 | 560 | ||
GenBank mRNA | BC061877.1 | - | 4 | 533 | 5599273 | |
CB578328.1 | - | 2 | 124 | |||
D00252.1 | - | 4 | 517 | |||
FM055803.1 | - | 4 | 482 | |||
FQ219727.1 | - | 1 | 48 | |||
J04171.1 | - | 4 | 501 |
