siRNA Details
| Gene Symbol: Grin2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012575.3 | NP_036707.3 | 14 | 3670 | ||
| XM_006247706.2 | XP_006247768.1 | 12 | 3389 | |||
| XM_006247707.1 | XP_006247769.1 | 15 | 3831 | |||
| XM_006247708.2 | XP_006247770.1 | 12 | 3286 | |||
| XM_006247709.2 | XP_006247771.1 | 12 | 3286 | |||
| GenBank mRNA | D13212.1 | - | 14 | 3670 | ||
| M91563.1 | - | 12 | 3289 | |||
| U08259.1 | - | 15 | 3830 | |||

Grin2c
Genomic Map