siRNA Details
| Gene Symbol: Grin2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012575.3 | NP_036707.3 | 4, 5 | 1400 | ||
| XM_006247706.2 | XP_006247768.1 | 2, 3 | 1119 | |||
| XM_006247707.1 | XP_006247769.1 | 5, 6 | 1561 | |||
| XM_006247708.2 | XP_006247770.1 | 2, 3 | 1016 | |||
| XM_006247709.2 | XP_006247771.1 | 2, 3 | 1016 | |||
| GenBank mRNA | D13212.1 | - | 4, 5 | 1400 | ||
| M91563.1 | - | 2, 3 | 1019 | |||
| U08259.1 | - | 5, 6 | 1560 | |||

Grin2c
Genomic Map