siRNA Details
| Gene Symbol: Grm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114330.1 | NP_001107802.1 | 5 | 1777 | ||
| NM_017011.1 | NP_058707.1 | 5 | 1777 | |||
| GenBank mRNA | M61099.1 | - | 5 | 1777 | ||
| X57569.1 | - | 5 | 1759 | |||
More Information
Set Membership: |
|

Grm1
Genomic Map