siRNA Details
Gene Symbol: Gusb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017015.2 | NP_058711.2 | 7 | 1207 | ||
GenBank mRNA | AY321342.1 | - | 1352 | |||
M13962.1 | - | 7 | 1177 | |||
Y00717.1 | - | 7 | 1133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gusb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017015.2 | NP_058711.2 | 7 | 1207 | ||
GenBank mRNA | AY321342.1 | - | 1352 | |||
M13962.1 | - | 7 | 1177 | |||
Y00717.1 | - | 7 | 1133 |