siRNA Details
| Gene Symbol: Hoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012581.2 | NP_036713.2 | 2 | 945 | ||
| GenBank mRNA | M91802.1 | - | 2 | 982 | ||
| Gene Symbol: NEWGENE_2813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762936.1 | XP_008761158.1 | 2 | 1526 | ||

Hoxa2
Genomic Map