siRNA Details
| Gene Symbol: Hoxc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109884.1 | NP_001103354.1 | 1 | 615 | ||
| XM_008765667.1 | XP_008763889.1 | 2 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc4| Gene Symbol: Hoxc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109884.1 | NP_001103354.1 | 1 | 615 | ||
| XM_008765667.1 | XP_008763889.1 | 2 | 1445 | |||