siRNA Details
| Gene Symbol: Hp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012582.2 | NP_036714.2 | 5 | 983 | ||
| GenBank mRNA | AF476963.1 | - | 5 | 1014 | ||
| AY325231.1 | - | 6 | 1111 | |||
| BC089816.1 | - | 5 | 983 | 7321960 | ||
| FQ209575.1 | - | 3 | 888 | |||
| FQ209592.1 | - | 5 | 1035 | |||
| FQ209684.1 | - | 5 | 1035 | |||
| FQ211144.1 | - | 5 | 1037 | |||
| FQ219122.1 | - | 5 | 1035 | |||
| FQ219443.1 | - | 5 | 1036 | |||
| FQ219547.1 | - | 5 | 1006 | |||
| FQ219598.1 | - | 5 | 1005 | |||
| K01933.1 | - | 2 | 747 | |||

Hp
Genomic Map