siRNA Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012583.2 | NP_036715.1 | 3 | 348 | ||
| GenBank mRNA | BC098629.1 | - | 3 | 348 | 7386548 | |
| M63983.1 | - | 3 | 380 | |||
| M86443.1 | - | 108 | ||||
| S79292.1 | - | 3 | 380 | |||
| X62085.1 | - | 308 | ||||
| Gene Symbol: LOC103689983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773659.1 | XP_008771881.1 | 3 | 424 | ||

Hprt1
Genomic Map