siRNA Details
| Gene Symbol: Il3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031513.1 | NP_113701.1 | 2, 3 | 248 | ||
| XM_006246226.2 | XP_006246288.1 | 2, 3 | 224 | |||
| GenBank mRNA | U81492.1 | - | 2, 3 | 248 | ||
| U81493.1 | - | 2, 3 | 239 | |||

Il3
Genomic Map