siRNA Details
| Gene Symbol: Il3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031513.1 | NP_113701.1 | 1 | 157 | ||
| XM_006246226.2 | XP_006246288.1 | 1 | 142 | |||
| GenBank mRNA | U81492.1 | - | 1 | 157 | ||
| U81493.1 | - | 1 | 157 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il3| Gene Symbol: Il3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031513.1 | NP_113701.1 | 1 | 157 | ||
| XM_006246226.2 | XP_006246288.1 | 1 | 142 | |||
| GenBank mRNA | U81492.1 | - | 1 | 157 | ||
| U81493.1 | - | 1 | 157 | |||