siRNA Details
| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017021.1 | NP_058717.1 | 5 | 458 | ||
| XM_006245901.2 | XP_006245963.1 | 4 | 800 | |||
| XM_006245903.2 | XP_006245965.1 | 5 | 820 | |||
| GenBank mRNA | L36459.1 | - | 5 | 458 | ||

Il9r
Genomic Map