siRNA Details
| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017021.1 | NP_058717.1 | 4 | 272 | ||
| GenBank mRNA | L36459.1 | - | 4 | 272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il9r| Gene Symbol: Il9r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017021.1 | NP_058717.1 | 4 | 272 | ||
| GenBank mRNA | L36459.1 | - | 4 | 272 | ||