siRNA Details
| Gene Symbol: Ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012592.2 | NP_036724.1 | 7 | 758 | ||
| GenBank mRNA | BC088401.1 | - | 7 | 758 | 7193077 | |
| FQ231497.1 | - | 6 | 600 | |||
| J05031.1 | - | 7 | 768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ivd| Gene Symbol: Ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012592.2 | NP_036724.1 | 7 | 758 | ||
| GenBank mRNA | BC088401.1 | - | 7 | 758 | 7193077 | |
| FQ231497.1 | - | 6 | 600 | |||
| J05031.1 | - | 7 | 768 | |||