siRNA Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052798.2 | NP_434685.1 | 4 | 1905 | ||
| XM_006246227.2 | XP_006246289.1 | 5 | 3052 | |||
| XM_008767660.1 | XP_008765882.1 | 5 | 3049 | |||
| XM_008767661.1 | XP_008765883.1 | 4 | 2953 | |||
| GenBank mRNA | BC078777.1 | - | 5 | 1739 | 7127868 | |
| M96548.1 | - | 4 | 1907 | |||

Zfp354a
Genomic Map