siRNA Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022667.2 | NP_073158.2 | 8 | 1043 | ||
| XM_006243655.1 | XP_006243717.1 | 9 | 1025 | |||
| GenBank mRNA | M64862.1 | - | 8 | 1044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco2a1| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022667.2 | NP_073158.2 | 8 | 1043 | ||
| XM_006243655.1 | XP_006243717.1 | 9 | 1025 | |||
| GenBank mRNA | M64862.1 | - | 8 | 1044 | ||