siRNA Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033235.1 | NP_150238.1 | 8 | 857 | ||
| GenBank mRNA | AF075574.1 | - | 2 | 71 | ||
| AF093773.1 | - | 8 | 857 | |||
| FQ212702.1 | - | 8 | 901 | |||
| FQ214669.1 | - | 3 | 196 | |||
| FQ215129.1 | - | 8 | 912 | |||
| FQ215603.1 | - | 8 | 896 | |||
| FQ217383.1 | - | 8 | 901 | |||
| FQ217819.1 | - | 901 | ||||
| FQ224891.1 | - | 8 | 899 | |||
| FQ233582.1 | - | 8 | 901 | |||

Mdh1
Genomic Map