siRNA Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033235.1 | NP_150238.1 | 2 | 113 | ||
| GenBank mRNA | AF093773.1 | - | 2 | 113 | ||
| BC059124.1 | - | 2 | 154 | 6921518 | ||
| FQ212702.1 | - | 2 | 157 | |||
| FQ215129.1 | - | 2 | 167 | |||
| FQ215603.1 | - | 2 | 152 | |||
| FQ217383.1 | - | 2 | 157 | |||
| FQ217819.1 | - | 157 | ||||
| FQ224891.1 | - | 2 | 155 | |||

Mdh1
Genomic Map