siRNA Details
| Gene Symbol: Me1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012600.2 | NP_036732.2 | 5, 6 | 610 | ||
| XM_008766439.1 | XP_008764661.1 | 5, 6 | 907 | |||
| GenBank mRNA | CO560778.1 | - | 1, 2 | 107 | ||
| FQ228526.1 | - | 5, 6 | 641 | |||

Me1
Genomic Map